BAH500250310
SP Bel-Art® Glove Box Bellows Type Gloves for 8 in. Glove Ports
latex, size 10 large
Synonym(s):
glove box
All Photos(1)
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
Recommended Products
material
latex
size
10 large
feature
to hold Bellows and Gloves
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art H50025-0310
General description
Made from natural latex with a removable hand, the sleeves fit 20cm (8") circular ports and 18 x 25cm (7 x 10") oval ports. Use with H50025-0320 Clamping Rings to hold gloves in place.
Features and Benefits
- Forms a leak-proof unit with O-ring construction integral with the box
- Bellows allows cooling air circulation
- When the glove and sleeve are separated, the sleeve remains attached to the box
- Glove sizes are interchangeable, and almost any standard laboratory, surgical or disposable glove can be used
- Length of the extended sleeve and glove is approximately 76cm (30")
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)