Skip to Content
Merck
All Photos(1)

Key Documents

900843

Sigma-Aldrich

Methanol-OD

reagent grade, ≥99 atom % D

Synonym(s):

mono-Deuteromethanol, Methan(ol-d), Methyl alcohol-OD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OD
CAS Number:
Molecular Weight:
33.05
Beilstein:
1730948
EC Number:
MDL number:
UNSPSC Code:
12142201

grade

reagent grade

isotopic purity

≥99 atom % D

Assay

≥99%

form

liquid

refractive index

n20/D 1.327 (lit.)

bp

65.5 °C (lit.)

density

0.813 g/mL at 25 °C (lit.)

mass shift

M+1

SMILES string

[2H]OC

InChI

1S/CH4O/c1-2/h2H,1H3/i2D

InChI key

OKKJLVBELUTLKV-VMNATFBRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service