Skip to Content
Merck
All Photos(1)

Key Documents

588571

Sigma-Aldrich

Butyric-3,3,4,4,4-d5 acid sodium salt

98 atom % (D)

Synonym(s):

Sodium butyrate-3,3,4,4,4-d5

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CD2CH2COONa
Molecular Weight:
115.12
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:

isotopic purity

98 atom % (D)

form

solid

mol wt

115.02 by atom % calculation

mp

250-253 °C (lit.)

mass shift

M+5

InChI

1S/C4H8O2/c1-2-3-4(5)6/h2-3H2,1H3,(H,5,6)/i1D3,2D2

InChI key

FERIUCNNQQJTOY-ZBJDZAJPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service