Skip to Content
Merck
All Photos(1)

Key Documents

232637

Sigma-Aldrich

1,3-Diphenoxybenzene

≥99%

Synonym(s):

Resorcinol diphenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OC6H4OC6H5
CAS Number:
Molecular Weight:
262.30
Beilstein:
2054179
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99%

form

powder

mp

59-61 °C (lit.)

SMILES string

O(c1ccccc1)c2cccc(Oc3ccccc3)c2

InChI

1S/C18H14O2/c1-3-8-15(9-4-1)19-17-12-7-13-18(14-17)20-16-10-5-2-6-11-16/h1-14H

InChI key

JTNRGGLCSLZOOQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

1,3-Diphenoxybenzene was used:
  • as internal standard during solid-phase extraction and gas chromatography coupled mass spectrometry in the selective ion monitoring (SIM) mode for the multi-class determination of pesticides
  • in the synthesis of new carboxylate ligands for simulation of binuclear active centres of nonheme oxygenases

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Van Hoof et al.
Journal of AOAC International, 84(5), 1420-1429 (2001-10-17)
As part of a project funded by the European Commission (EC) for the development and evaluation of multiresidue methods for analysis of drinking and related waters, 15 European laboratories evaluated a method using styrene-divinylbenzene co-polymer solid-phase extraction followed by gas
Synthesis of new carboxylate ligands for simulation of binuclear active centers of nonheme oxygenases.
Gumanov LL, et al.
Russ. J. Org. Chem., 38(4), 606-608 (2002)
Martin Lommatzsch et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 33(3), 473-488 (2016-01-09)
Hot-melt adhesives are widely utilised to glue cardboard boxes used as food packaging material. They have to comply with the requirements of Article 3 of the European Framework Regulation for food contact materials (1935/2004). The hot melt raw materials analysed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service