Merck
Dichotomy between the T and the B cell epitopes of the synthetic polypeptide (T,G)-A--L.
European journal of immunology (1994-10-01)
E Zisman, M Sela, A Ben-Nun, E Mozes
RÉSUMÉ
Studies with the well-characterized, synthetic, random-multichain polypeptide poly(LTyr,LGlu)-poly(DLAla)-poly(LLys) (T,G)-A-L) led to the discovery of determinant-specific genetic control of the immune response, as well as to other immunological phenomena. Moreover, the tetrapeptide TyrTyrGluGlu built on the same backbone ("(T-T-G-G)-A--L") was found to represent its major B cell epitope. We have recently shown that for interaction with major histocompatibility complex class II molecules and stimulation of T cells, (T,G)-A--L requires proteolytic processing and the resulting T cell epitopes are close to the N termini of the branched polymer's side chains. Thus, we were interested to elucidate the major T cell epitope of (T,G)-A--L, by using the ordered polypeptides (T-T-G-G)-A--L and (T-G-T-G)-A--L, in which only the two internal amino acids of the tetrapeptide attached to the side chains are switched. We established T cell lines to these antigens, and found that the ordered analog (T-T-G-G-)-A--L, which was defined as the B cell epitope of (T,G)-A--L, did not represent its T cell epitope, whereas (T-G-T-G)-A--L, to which only a minor anti-(T,G)-A--L Ab response was directed, was found to be its major T cell epitope. In addition, there was no cross-reaction between (T-G-T-G)-A--L and (T-T-G-G)-A--L at the T cell level, similar to the lack of cross-reaction of their antibodies. Analysis of the repertoire of the T cell receptors used by these lines revealed that the (T,G)-A--L and the (T-T-G-G)-A--L specific T cell lines were not restricted in their V alpha and V beta TCR usage, whereas the (T-G-T-G)-A--L-specific line was restricted by both V alpha and V beta T cell receptor gene products. This difference might be due to the thymus-independent characteristics previously described for the latter antigen.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Poly(Ala, Glu, Lys, Tyr) 6:2:5:1 hydrobromide, Ala:Glu:Lys:Tyr (6:2:5:1), mol wt 20,000-30,000