Merck
Identification of Ginkgo biloba as a novel activator of pregnane X receptor.
Drug metabolism and disposition: the biological fate of chemicals (2008-08-30)
Eugene Y H Yeung, Tatsuya Sueyoshi, Masahiko Negishi, Thomas K H Chang
RÉSUMÉ
Pregnane X receptor (PXR; NR1I2) is a ligand-activated transcription factor that plays a role not only in drug metabolism and transport but also in various other biological processes. Ginkgo biloba is a herbal medicine commonly used to manage memory impairment. Treatment of primary cultures of rat hepatocytes with G. biloba extract increases the mRNA expression of CYP3A23, which is a target gene for rat PXR. The present study was conducted to test the hypothesis that G. biloba extract activates PXR. Treatment of mouse PXR (mPXR) or human PXR (hPXR)-transfected HepG2 cells with G. biloba extract at 200 microg/ml increased mPXR and hPXR activation by 3.2- and 9.5-fold, respectively. Dose-response analysis showed a log-linear increase in hPXR activation by the extract over the range of 200 to 800 microg/ml. To determine whether G. biloba extract induces hPXR target gene expression, cultured LS180 human colon adenocarcinoma cells were treated for 72 h with the extract. G. biloba extract at 200, 400, and 800 microg/ml increased CYP3A4 mRNA expression by 1.7-, 2.4-, and 2.5-fold, respectively. The same concentrations of the extract increased CYP3A5 (1.3-3.6-fold) and P-glycoprotein (ABCB) 1 (2.7-6.4-fold) mRNA expression. At concentrations (5 and 10 microM) that did not down-regulate PXR gene expression and were not cytotoxic, L-sulforaphane (an hPXR antagonist) decreased CYP3A4, CYP3A5, and ABCB1 gene expression in cells treated with G. biloba extract. In summary, G. biloba extract activated mPXR and hPXR in a cell-based reporter gene assay and induced CYP3A4, CYP3A5, and ABCB1 gene expression in hPXR-expressing LS180 cells.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
L-Sulforaphane, ≥95% (HPLC), oil