Merck
Nucleosome eviction and activated transcription require p300 acetylation of histone H3 lysine 14.
Proceedings of the National Academy of Sciences of the United States of America (2010-10-27)
Whitney R Luebben, Neelam Sharma, Jennifer K Nyborg
RÉSUMÉ
Histone posttranslational modifications and chromatin dynamics are inextricably linked to eukaryotic gene expression. Among the many modifications that have been characterized, histone tail acetylation is most strongly correlated with transcriptional activation. In Metazoa, promoters of transcriptionally active genes are generally devoid of physically repressive nucleosomes, consistent with the contemporaneous binding of the large RNA polymerase II transcription machinery. The histone acetyltransferase p300 is also detected at active gene promoters, flanked by regions of histone hyperacetylation. Although the correlation between histone tail acetylation and gene activation is firmly established, the mechanisms by which acetylation facilitates this fundamental biological process remain poorly understood. To explore the role of acetylation in nucleosome dynamics, we utilized an immobilized template carrying a natural promoter reconstituted with various combinations of wild-type and mutant histones. We find that the histone H3 N-terminal tail is indispensable for activator, p300, and acetyl-CoA-dependent nucleosome eviction mediated by the histone chaperone Nap1. Significantly, we identify H3 lysine 14 as the essential p300 acetylation substrate required for dissociation of the histone octamer from the promoter DNA. Together, a total of 11 unique mutant octamer sets corroborated these observations and revealed a striking correlation between nucleosome eviction and strong activator and acetyl-CoA-dependent transcriptional activation. These novel findings uncover an exclusive role for H3 lysine 14 acetylation in facilitating the ATP-independent and transcription-independent disassembly of promoter nucleosomes by Nap1. Furthermore, these studies directly couple nucleosome disassembly with strong, activator-dependent transcription.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Sigma-Aldrich
Acétyl-coenzyme A trisodium salt, ≥93% (HPLC), powder
Sigma-Aldrich
Acétyl-coenzyme A lithium salt, ≥93% (HPLC)