Principaux documents
58768
Coupleur
1/4-28 for 1/16 in. O.D., bore diam. 0.043 in.
Synonyme(s) :
Raccords SSI
About This Item
Code UNSPSC :
41115700
Matériaux
stainless steel
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
SSI
Diam. de l'orifice
0.043 in.
Raccords
1/4-28 for 1/16 in. O.D.
Description générale
Acier inoxydable. Filetages 1/4-28
Produits recommandés
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Informations légales
SSI is a trademark of Scientific Systems, Inc.