Principaux documents
2637105
Supelco
Insert d'injection droit, sans division (non rempli)
pkg of 5 ea
About This Item
Code UNSPSC :
41115719
Description
Split (unpacked)
Conditionnement
pkg of 5 ea
L × D.E. × D.I.
54 mm × 5.0 mm × 3.4 mm
Description générale
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Varian (1078 and 1079 Injector)