Documents
24808-U
Supelco
Supeltex M-4 Capillary Ferrule, Short Design
I.D. 0.8 mm, 0.53 mm Column I.D., pkg of 10 ea
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
SB.54
Caractéristiques
ferrule type short
Conditionnement
pkg of 10 ea
Fabricant/nom de marque
Agilent/HP 500-2118 (OEM)
D.I.
0.8 mm
Compatibilité
0.53 mm Column I.D.
Description générale
Swage type fittings are one of the most popular connectors which are available in metal and plastic forms; they are very useful for connecting similar or dissimilar materials, like metal to glass or metal to plastic. The general swage tubing coupler has four parts namely the body, into which the tube is placed, two ferrules that is front ferrule and back ferrule and lastly a packing nut. If one is using the original nut supplied for the injector and non-MS detector end of an GC instrument, one should choose a Supeltex short design ferrule. Supeltex M-4 (flexible graphite) ferrules provide great sealability, ensuring a leaktight seal for extended periods at high temperature. It has limited reusability.
Supelco a amélioré le dessin de ces ferrules en graphite pour obtenir un profil propre et net, avec un flash minimum.
Caractéristiques :
devient étanche après serrage manuel d′un quart de tour. Contact maximum de la surface d′étanchéité, risque moindre de contamination de la colonne lors de l′installation.
Composition du matériau : graphite souple
Température maximum : 450 °C
Informations légales
Configuré(e)(s) pour
Réf. du produit
Description
Tarif
Choose from one of the most recent versions:
Certificats d'analyse (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Service Clients
Déjà en possession de ce produit ?
The Manipulation of Air-Sensitive Compounds.
Shriver DF.
Nature, 217-218 (1986)