Principaux documents
21642
Supelco
Agilent® 5880, 5890, 5987, 6890 (S23965)
L × O.D. × I.D. 10.0 ft (3.0 m) × 1/4 in. × 2.0 mm
About This Item
Code UNSPSC :
41121800
Matériaux
glass column
Technique(s)
gas chromatography (GC): suitable
L × D.E. × D.I.
10.0 ft (3.0 m) × 1/4 in. × 2.0 mm
Type de colonne
empty
Compatibilité
for use with Agilent Configuration B
Description générale
GC columns are of two types; packed column and capillary column. Capillary column carries the gas through a central aperture which is unimpeded through the entire length of column. In general, capillary columns are made longer to produce more theoretical plates. This also helps in band dispersion eventually narrowing the peaks and resulting in high resolution.
Silane treated empty glass column. Fittings are not included.
  • X (length of injector arm) = 11.02 in. (280 mm)
  • Y (length of detector arm) = 7.09 in. (180 mm)
  • S (span, injector to detector) = 9.0 in. (229 mm)
This column fits Agilent 5880, 5890, 6890, and 7890 (configuration B, on-column injection, TCD only).
Application
Empty column may be used for packing adsorbents for temperature-programmed desorption (TPD) instrumentation.
Informations légales
Agilent is a registered trademark of Agilent Technologies, Inc.
The determination of active filter aid adsorption sites by temperature-programmed desorption.
Zhu. Z. Y.
Journal of the American Oil Chemists' Society, 71, 189-194 (1994)
Raymond P. W. Scott
Introduction to Analytical Gas Chromatography (1997)