Principaux documents
Z763586
Incu-Shaker universal spring platform
L × W 14 in. × 12 in.
Synonyme(s) :
shaker spring platform
1 EA
751,00 €
751,00 €
Check Cart for Availability
Changer de vue
1 EA
751,00 €
About This Item
Code UNSPSC :
41103811
Nomenclature NACRES :
NB.77
751,00 €
Check Cart for Availability
Fabricant/nom de marque
Benchmark Scientific H1010-P-SP
L × l
14 in. × 12 in.
Description générale
Universal platform for the Incu-Shaker.
Informations légales
Incu-Shaker is a trademark of Benchmark Scientific, Inc.