This is an ice pack for sample storage that provides an alternative to conventional ice. There is no information on using this for personal care.
Z763195
NEW ice, ice packs
pack of 45 ea
15,40 €
About This Item
15,40 €
Produits recommandés
Matériaux
polyethylene
Conditionnement
pack of 45 ea
Fabricant/nom de marque
Diversified Biotech NEW-100
Description générale
- FDA-approved
- Made of durable polyethylene
- Washable
- Re-useable, Re-freezable
- No watery mess to clean up
-
This is just an ice pack, correct? We want to use it prior to doing a facial procedure. Is this not allowed with your ice pack?
1 réponse-
Utile ?
-
-
Can you provide the dimensions of these ice packs or cubes?
1 réponse-
As per the vendor, the ice pack cubes are approximately 2 inches square.
Utile ?
-
-
Can I please know the dimensions of the gel packs?
1 réponse-
These items are approximately 2 inches square.
Utile ?