Principaux documents
Z743489
Aluminum heating/cooling block
for 384-well plates, sbs footprint
Synonyme(s) :
dry block, plate cooling block, plate heating block, tube cooling block, tube heating block
About This Item
Code UNSPSC :
41102404
Nomenclature NACRES :
NB.77
Caractéristiques
for 384-well plates, sbs footprint
Fabricant/nom de marque
Diversified Biotech CHAM-A384
Description générale
Each Cooling Chamber is a solid aluminum block that surrounds inserted tubes to guarantee sustained hot or cold temperatures.
Les clients ont également consulté
Slide 1 of 2
1 of 2