Principaux documents
Z742102
Cell Strainer
blue, Individually Wrapped, sterile; γ-irradiated, pack of 50 ea
About This Item
Code UNSPSC :
41104932
Nomenclature NACRES :
NB.22
Matériaux
Nylon
blue
Stérilité
sterile; γ-irradiated
Conditionnement
pack of 50 ea
pkg of (individually wrapped)
Fabricant/nom de marque
MIDSCI SG-40ICS
Dimension de pores
40 μm
Description générale
Cell Strainers from MIDSCI can be used for many applications including isolating cells and removing clumps or debris. Cell Strainers from MIDSCI are compatible with standard 50mL centrifuge tubes. The exclusive Pestle design improves manipulation of material in the strainer during operation.
  • Polypropylene housing with nylon mesh filter eliminates the need to fabricate filters from gauze, paper, nylon mesh, or other materials
  • Elongated lip on housing extends beyond edge of centrifuge tube enabling aseptic handling with forceps
  • Extended gripping tab on housing for easy handling
  • Available in three porosities: 40, 70, 100 um
  • Color coded by mesh porosity for easy identification
  • Packaged sterile (gamma irradiated) in individually wrapped peel-to-open paper/plastic wrapper or bulk packed in bags
Jessica M Vélez et al.
PloS one, 16(1), e0231367-e0231367 (2021-01-07)
The ectomycorrhizal fungal symbiont Cenococcum geophilum is of high interest as it is globally distributed, associates with many plant species, and has resistance to multiple environmental stressors. C. geophilum is only known from asexual states but is often considered a
Articles
Take a deep dive into the world of 3D cell culture courtesy of Corning and understand why the Corning® spheroid microplate can be an asset in the evolution of 3D cell culture.
Protocoles
Understand how Corning® Spheroid Microplates can be used for generating and culturing tumor spheroids in a 96-well ­spheroid microplate format.
Step-by-step tumorpshere formation protocol that can used to generate 3D cancer stem cell spheroids.