Principaux documents
Z708410
Refrigerator rack
Holds 50 x 15 or 50 mL tubes
About This Item
Code UNSPSC :
41122806
Nomenclature NACRES :
NB.22
Matériaux
polystyrene
Caractéristiques
Holds 50 x 15 or 50 mL tubes
Fabricant/nom de marque
Heathrow Scientific HS28230B
Technique(s)
cryopreservation: suitable
Taille
276x164x134 mm
Description générale
Quick chill sample tubes. Two-piece rack has a lower section tray which can hold water or crushed ice. Top insert tray secures tubes. Molded polystyrene racks are designed to stack on top of each other to save benchtop space.
Les clients ont également consulté
Slide 1 of 1