Principaux documents
Z659339
Bandelin SONOPULS ultrasonic homogenizer accessories
Flow-through vessel DG 5 for HD 2200 homogenizer
Synonyme(s) :
sonification vessel
1 EA
388,00 €
388,00 €
Check Cart for Availability
Changer de vue
1 EA
388,00 €
About This Item
Code UNSPSC :
41101520
Nomenclature NACRES :
NB.74
388,00 €
Check Cart for Availability
Matériaux
glass
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bandelin 482
Disponibilité
available only in EU
Volume × D.I. × H
80 mL × 35 mm × 95 mm
Description générale
Flow vessel DG with cooling jacket, for continuous sonication of larger quantities in the flow. The cooling jacket allows temperature control using liquid coolants during sonication via existing hose connections.
Caractéristiques et avantages
  • Simultaneous sound and cooling
  • Optimal process observation