Z640557
Eppendorf® epT.I.P.S. Biopur®, racks
volume 0.1-20 μL
Synonyme(s) :
pipetting tips
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.01
Produits recommandés
Matériaux
colorless
Stérilité
non-sterile
Conditionnement
pack of 480 (5 racks of 96 tips)
Fabricant/nom de marque
Eppendorf® 0030 075.005
Eppendorf® 022491067 (USA)
Plage de mesure du volume
0.1-20 μL
Longueur
40 mm
Volume maximum
20 μL
Description générale
Biopur guaranteed sterile, pyrogen-free, RNase-free, DNA-free and ATP-free, for applications in pharmaceutical and food industries, molecular biology and cell technology.
Batch specific certificates available on the internet at http://www.eppendorf.com
Batch specific certificates available on the internet at http://www.eppendorf.com
Informations légales
Biopur is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG