Z363723
Transilluminator, UV, accessory
Replacement Bulb, Midrange UV, fluorescence λem 312 nm
About This Item
Code UNSPSC :
41105300
Produits recommandés
Fluorescence
λem 312 nm
Autres remarques
To replace bulbs shipped with UV transilluminators