Z332607
BRAND® HandyStep® repetitive pipette
About This Item
Code UNSPSC :
41121500
Description générale
Brand HandyStep works on the positive displacement principle, making it ideal for routine dispensing as well as for difficult liquids. Viscous, high density or high vapor pressure liquids can be dispensed while maintaining accuracy and precision. Optimal results are obtained with PLASTIBRAND PD-Tips. The HandyStep is also compatible with Combitips plus, Combitips and other standard repetitive tips. Conformity certified, with serial number and performance certificate, CE-marked according to IVD-Directive 98/79 CE.
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG
HandyStep is a registered trademark of BRAND GMBH + CO KG