Documents
Z326364
Sigma-Aldrich
BRAND® SILBERBRAND volumetric flask
volume 2,000 mL, accuracy: 0.9 mL
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
Joint
joint ST/NS neck29/32
Stérilité
non-sterile
Caractéristiques
class B DIN EN ISO 1042, calibrated to contain (TC, IN)
closure type PP Stopper
Conditionnement
pack of 1
Fabricant/nom de marque
BRAND
Plage de mesure du volume
2,000 mL, accuracy: 0.9 mL
Capacité
2000 mL
Description générale
Volumetric flasks according to DIN EN ISO 1042, Class B. Made from Boro 3.3, calibrated to contain (TC, IN). Supplied with a square grip PP stopper. Marks and inscriptions in high contrast white enamel.
Autres remarques
BR36754 (India)
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG