Principaux documents
Z317330
Eppendorf® racks for microcentrifuge tubes
For 24 × 1.5 ml - 2 ml tubes
About This Item
Code UNSPSC :
41103900
Fabricant/nom de marque
Eppendorf® 0030 123.115
Eppendorf® 022364227
Disponibilité
not available in USA and Canada
Description générale
  • 24-place capacity
  • Unique swivel connector to interlock multiple racks for front-to-back and side-to-side configurations
  • Non-slip stand
  • Numbered positions
  • UV-resistant
  • Autoclavable
Informations légales
Eppendorf is a registered trademark of Eppendorf AG