Principaux documents
TMO3110-0160
Nalgene® high-speed centrifuge tubes
round bottom natural polypropylene copolymer, volume 16 mL, without cap, non-sterile, case of 100 ea
Synonyme(s) :
Nalgene tubes, round bottom tubes
100 EACH
659,00 €
659,00 €
Date d'expédition estimée le12 mai 2025
Changer de vue
100 EACH
659,00 €
About This Item
Code UNSPSC :
41121703
Nomenclature NACRES :
NB.11
659,00 €
Date d'expédition estimée le12 mai 2025
Matériaux
round bottom natural polypropylene copolymer
Stérilité
non-sterile
Caractéristiques
without cap
Conditionnement
case of 100 ea
Fabricant/nom de marque
Nalgene 3110-0160
Volume
16 mL
Description générale
Improve rotor fit and performance using Thermo Scientific Nalgene High-Speed Round-Bottom PPCO Centrifuge Tubes, featuring uniform wall thickness and diameter. Tubes have excellent chemical resistance.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries