Principaux documents
OH83033968
Ohaus® pH Electrode
ST230
About This Item
Code UNSPSC :
41115604
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Ohaus® 83033968
Plage de mesure du pH
0.00-14 pH unit
Description générale
All electrodes include a temperature sensor, which powers automatic temperature compensation and ensures accurate measurements.
Application
pH, reference, oxidation-reduction potential (ORP), conductivity, dissolved oxygen (DO), temperature measurement
Caractéristiques et avantages
Can be used in conjunction with all Starter bench and portable meters
Informations légales
Ohaus is a registered trademark of Ohaus Corp.