Principaux documents
HS8881A
Vinyl-Coated Lead Ring ("C" shape)
fits beaker size, 125 to 500 mL, blue
1 EACH
34,10 €
34,10 €
Check Cart for Availability
Changer de vue
1 EACH
34,10 €
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
34,10 €
Check Cart for Availability
Matériaux
blue
Caractéristiques
fits beaker size 125 to 500 mL
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Heathrow Scientific HS8881A
Description générale
The Lead Ring is perfect for stabilizing vessels and labware or for water bath immersions.
Caractéristiques et avantages
  • Vinyl-coated lead ring will not mark or scratch benchtops and labware
  • O-Shape Style