Principaux documents
GN780275
Greiner MASTERBLOCK® storage microplate
96 well, polypropylene, capacity 2.0 mL, conical (V) bottom, green
About This Item
Code UNSPSC :
41121800
Matériaux
polypropylene
Conditionnement
pack of 50 ea
Fabricant/nom de marque
Greiner 780275
Disponibilité
available only in China
Taille
96 wells
Adéquation
suitable for (general storage, liquid handling; automated applications)
Informations légales
MASTERBLOCK is a registered trademark of Greiner Bio-One GmbH