Documents
GE28-4064-13
Sigma-Aldrich
Tricorn 10/20 Column
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
SB.52
Technique(s)
LPLC: suitable
Catégories apparentées
Description générale
Tricorn 5 and 10 Coarse Filter Kit is used for large volume/repeated loading of clarified sample to reduce the risk of clogging in combination with capture media. Both filter and coarse filter kits include top and bottom adapters, bottom filters, and EPDM O-rings, five of each.
Tricorn Packing Equipment includes the Packing Equipment 5/50 (5-mm connector and 50-mm glass tube) or 10/100 (10-mm connector and 100-mm glass tube), EPDM O-rings and bottom unit, cap, and stop plug.
Caractéristiques et avantages
Broad range of column dimensions suitable for many different purification needs.
Glass tubes coated with plastic film prevent harm if the column is dropped or operated outside its specification.
Locking ring prevents accidental compression of media bed.
Separate packing connector allows simple column packing for a broad range of media.
The modern integrated, snap-on design results in a column that is easy to assembly, operate, and maintain.
Tricorn empty columns are designed for high performance separations on small and medium scale. Pressure stability up to 100 bar and durable column material ensure reliable and reproducible operation as well as compatibility with the majority of the commercial lab media.
Articles
This page describes efficient column packing and preparation for affinity chromatography of antibodies.
Desalting at laboratory scale is a well-proven, simple, and fast method that will rapidly remove low molecular weight contaminants at the same time as transferring the sample into the desired buffer in a single step.
This page shows how to solve practical problems that may occur when running an affinity chromatography column.
Protocoles
This page describes efficient column packing and preparation for affinity chromatography of GST-tagged proteins using Cytiva products.
This page shows how to purify histidine-tagged proteins secreted into eukaryotic cell culture supernatants using Ni Sepharose Excel from Cytiva.
Column Packing and Preparation for Size Exclusion Chromatography
This page describes efficient column packing and preparation for affinity chromatography of tagged proteins using Cytiva products.
Afficher tout