EP5804712005
Eppendorf® Aerosol-tight caps
for 100 ml rectangular buckets, set of 2 ea
134,00 €
About This Item
134,00 €
Produits recommandés
Caractéristiques
for 100 ml rectangular buckets
Conditionnement
set of 2 ea
Fabricant/nom de marque
Eppendorf® 5804712005
Technique(s)
centrifugation: suitable
Compatibilité
for use with 5804 R
for use with 5804
for use with 5810 R
for use with 5810