CLS9016
Corning® aspiration pipettes
volume 2 mL, polystyrene, plain, paper/plastic wrapped
Synonyme(s) :
Aspiration Pipettes, Corning Pipettes
326,00 €
About This Item
326,00 €
Produits recommandés
Matériaux
polystyrene
Stérilité
sterile
Conditionnement
pack of 50 ea
case of 500 ea
Fabricant/nom de marque
Corning 9016
Capacité
2 mL
Volume
2 mL
Catégories apparentées
Description générale
- Corning® aspirating pipettes are made of optically clearpolystyrene for durability and good mechanical strength.
- The sterile,ungraduated, and unplugged pipettes are best suited for aspirating liquidaseptically using vacuum suction.
- Individually wrappedand bulk packed.
- Disposable andcertified non-pyrogenic.