Principaux documents
CLS64061L
PYREX® economy Squibb separatory funnel
polyethylene stopper, PTFE stopcock, pack of 1 ea
Synonyme(s) :
pouring funnel
About This Item
Code UNSPSC :
41121809
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
PTFE stopcock
polyethylene stopper
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Corning 6406-1L
Corps, H × D.E.
395 mm × 119 mm
Capacité
1 L
Tige, L × D.E.
52 mm × 7 mm
Volume
1 L
1000 mL
Catégories apparentées
Description générale
These economical separatory funnels are designed to reduce replacement costs by providing replaceable components. The sturdy, durable funnels are designed for long life and maximum safety. The replaceable stopcock assembly has a simple screw thread locking nut and collar which ensure that the stopcock cannot fall out in use. The tip has a 7mm O.D. stem that is also replaceable. Its ground end provides a better grip and helps prevent the tip from falling out.
Informations légales
PYREX is a registered trademark of Corning, Inc.