Principaux documents
CLS4519167
Corning® ProCulture® spinner flask accessories
Gas and Media Handling Fitting with Triple 1/8 in. Inlets (2 long, 1 short tube) for 3L Angled Sidearm
Synonyme(s) :
container accessory
1 EA
630,00 €
630,00 €
Check Cart for Availability
Changer de vue
1 EA
630,00 €
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
630,00 €
Check Cart for Availability
Fabricant/nom de marque
Corning 4519-167
Catégories apparentées
Description générale
Side arm fittings used for a variety of functions in angled sidearm Spinner Flasks during cell culture. Sidearm fittings provide a convenient way of transferring media into or out of flasks aseptically. They can also be used to sparge the cell culture medium with gases to enhance cell production. Fittings comprise of a PET insert with a Viton O-ring and a polypropylene sealing cap & 316 stainless steel tubes.
Dual tubes extend to the bottom of the flask, and are held in place by Noryl nuts with integrated ferrules.
Combination tubes has two different length tubes, one extends to the bottom of the flask; the other is a shorter 6′ length. Both tubes are held in place by Noryl nuts with integrated ferrules.
Informations légales
Corning is a registered trademark of Corning, Inc.
ProCulture is a registered trademark of SP Industries