Principaux documents
CLS3507
Corning® 96 well filter plates
case of 1, pack of 11 ea
Synonyme(s) :
96 multiwell plate
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.11
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Nom du produit
Corning® 96 well filter plate, volume adapter applicator, 96 well filter plate volume adapter applicator, for 96 well filter plates, 1/cs
Conditionnement
case of 1
pack of 1
Fabricant/nom de marque
Corning 3507
Adéquation
suitable for (96 well filter plates)
Description générale
Corning 96 Well Filter Plate, Volume Adapter Applicator
The Corning 96 well filter plate volume adaptor applicator easily assembles and disassembles the filter plate and adapter and ensures a perfect, leak-free fit.
Informations légales
Corning is a registered trademark of Corning, Inc.