Accéder au contenu
Merck
Toutes les photos(1)

Principaux documents

C213640

Sigma-Aldrich

DMT-2′O-TC-rC(ac) Phosphoramidite

configured for MerMade

Synonyme(s) :

N4-Acetyl-O5′-(4,4′-dimethoxytrityl)-O2′-(1,1-dioxothiomorpholine-4-thiocarbonyl)cytidine O3′-(O-(2-cyanoethyl)-N,N-diisopropylphosphoramidite)

Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme


About This Item

Formule empirique (notation de Hill):
C46H57N6O11PS2
Numéro CAS:
Poids moléculaire :
965.08
Numéro MDL:
Code UNSPSC :
12352200
ID de substance PubChem :
Nomenclature NACRES :
NA.51

Gamme de produits

Proligo Reagents

Essai

≥98% (31P-NMR)
≥98.0% (reversed phase HPLC)

Forme

solid

Technique(s)

oligo synthesis: suitable

Impuretés

≤0.3 wt. % water content (Karl Fischer)
≤0.5% 3′-TC isomer (reversed phase HPLC)
≤0.5% P(III) Impurities 145-155ppm (31P-NMR)

Couleur

white to light yellow

Solubilité

acetonitrile: toluene (1:1): 0.1 M at 18 °C, clear

λ

conforms (UV/VIS Identity)

Compatibilité

configured for MerMade

Profil des nucléosides

base: cytidine
base protecting group: acetyl
2' protecting group: TC
5' protecting group: DMT
deprotection: fast

Température de stockage

−20°C

Chaîne SMILES 

COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](OC(=S)N3CCS(=O)(=O)CC3)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)N4C=CC(NC(C)=O)=NC4=O)(c5ccccc5)c6ccc(OC)cc6

InChI

1S/C46H57N6O11PS2/c1-31(2)52(32(3)4)64(60-27-11-23-47)63-41-39(30-59-46(34-12-9-8-10-13-34,35-14-18-37(57-6)19-15-35)36-16-20-38(58-7)21-17-36)61-43(51-24-22-40(48-33(5)53)49-44(51)54)42(41)62-45(65)50-25-28-66(55,56)29-26-50/h8-10,12-22,24,31-32,39,41-43H,11,25-30H2,1-7H3,(H,48,49,53,54)/t39-,41-,42-,43-,64?/m1/s1

Clé InChI

OUGOPIOQANCAOY-JFALYFJISA-N

Vous recherchez des produits similaires ? Visite Guide de comparaison des produits

Informations légales

For use with Customer Container sold to end user customers either directly or through distributors.

This product is manufactured and sold under a license from Agilent Technologies under under US Patent No. 8,202,983 and pending U.S. Patent App. Nos. 13/485,5925, 13/531,191, and 13/339,082 and corresponding foreign counterparts, including those issuing from PCT/US08/63342 and PCT/US09/057922 all assigned to Agilent Technologies. The purchase of this product conveys to the buyer the limited, non-exclusive, non-transferable right (without the right to resell, repackage, or further sublicense) under these patents to use the product to synthesize Oligonucleotides for buyer′s internal research use only, including commercial purposes. Except as otherwise provided herein, no other license is granted to the buyer weather expressly, by implication, by estoppel or otherwise. Buyer agrees and understands that it is not licensed to use this product to synthesize Oligonucleotides for sale, for diagnostic uses including use in clinical diagnostic settings where the use in conjunction with an individual′s sample generates data provided to an individual or the individual′s caregiver for a free reimbursement, neither can Oligonucleotides synthesized using this product be used for use clinical trials or for other therapeutic purposes. This product is sold pursuant to authorization from Agilent Technologies, Inc. and Agilent Technologies reserves all other rights under these patents.

For use with all sales where the purchaser is required to have its own direct license from Agilent, without limitation, Bulk Quantity Sales, and sales to Oligo Houses.

This product and it uses are covered by under US Patent No. 8,202,983 and pending U.S. Patent App. Nos. 13/485,5925, 13/531,191, and 13/339,082 and corresponding foreign counterparts, including those issuing from PCT/US08/63342 and PCT/US09/057922 all assigned to Agilent Technologies. This product is sold to purchaser pursuant to authorization by Agilent Technologies, Inc. without any rights conferred by seller, based on purchaser′s direct license and authorization to purchase from Agilent Technologies, Inc. No license is granted to the purchaser weather expressly, by implication, by estoppel or otherwise on account of this purchase. Purchase′s rights to use this product are governed solely and expressly by purchaser′s direct license agreement with Agilent.
These products are manufactured and sold under a license from Agilent Technologies under US Patent No. 8,202,983 and pending U.S. Patent App. Nos. 13/485/5925, 13/531,191, and 13/339,082 and corresponding foreign counterparts, including those issuing from PCT/US08/63342 and PCT/US09/057922 all assigned to Agilent Technologies.

These products and its uses are covered by US Patent No. 8,202,983 and pending U.S. Patent App. Nos. 13/485/5925, 13/531,191, and 13/339,082 and corresponding foreign counterparts, including those issuing from PCT/US08/63342 and PCT/US09/057922 all assigned to Agilent Technologies.

Code de la classe de stockage

13 - Non Combustible Solids

Classe de danger pour l'eau (WGK)

WGK 3

Point d'éclair (°F)

Not applicable

Point d'éclair (°C)

Not applicable


Faites votre choix parmi les versions les plus récentes :

Certificats d'analyse (COA)

Lot/Batch Number

Vous ne trouvez pas la bonne version ?

Si vous avez besoin d'une version particulière, vous pouvez rechercher un certificat spécifique par le numéro de lot.

Déjà en possession de ce produit ?

Retrouvez la documentation relative aux produits que vous avez récemment achetés dans la Bibliothèque de documents.

Consulter la Bibliothèque de documents

Notre équipe de scientifiques dispose d'une expérience dans tous les secteurs de la recherche, notamment en sciences de la vie, science des matériaux, synthèse chimique, chromatographie, analyse et dans de nombreux autres domaines..

Contacter notre Service technique