Principaux documents
BMSW110521100
Sigma-Aldrich
Accuris M1 Grade Calibration Weight Set
pack of 21 pieces, size 0.001-100 g, stainless steel
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.71
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Matériaux
stainless steel
Conditionnement
pack of 21 pieces
Fabricant/nom de marque
Benchmark Scientific W1105-21100
Taille
0.001-100 g
Description générale
Basic, compact balances with readability in the range of 0.1g to 0.01g can be calibrated with M1 Class weights.
Each individual weight and weight set from Accuris is supplied with an individual Certificate of Verification confirming that the weight(s) meet OIML regulations and tolerances for the specific weight class.
The economy M1 Calibration Weight Set includes 21 pieces in a protective carrying case.
Informations légales
Accuris is a trademark of Benchmark Scientific, Inc.