Merck
Passer à
B8907
Nalgene® bottles, style 2104
capacity 60 mL
Synonyme(s) :
HDPE bottles, nalgene bottle, nalgene bottles
Se connecter pour consulter les tarifs organisationnels et contractuels.
A propos de cet article
NACRES:
NB.17
UNSPSC Code:
41121800
material
polypropylene cap, polypropylene screw closure, round bottle, translucent high-density polyethylene bottle
joint
Joints threaded neck (28)
description
Not autoclavable
feature
with cap
packaging
pack of 12 ea
manufacturer/tradename
Nalgene® 2104-0002
bottle diam. × H
39 mm × 86 mm
capacity
60 mL
General description
Boston-style (round) wide-mouth bottles
Features and Benefits
  • Nalgene® bottles are ideal for storing chemicals, materials, and specimens.
  • Laboratory-grade HDPE material for reliably low leachables and extractables.
  • Suitable for use in freezers (up to -100°C).
  • Rigid, uniform walls offer protection against splitting or punctures.
  • Bottles have excellent durability and are chemically resistant to most acids, bases, and alcohols.
  • Linerless caps ensure a leak-proof seal for sample protection.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries