Documents
AXYVTF38450LRS
Corning® Axygen® Automation Tips for Agilent/["Velocity11 VPrep"] and Bravo
volume (50 μL), Filtered, non-sterile, Maxymum Recovery
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.02
Stérilité
non-sterile
Conditionnement
case of 19,200 tips
pack of 3,840 tips
pack of 10 racks
case of 5 packs
pkg of racked
Fabricant/nom de marque
Corning VTF-384-50-L-R-S
Volume
(50 μL)
Informations légales
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.