Accéder au contenu
Merck
Toutes les photos(1)

Principaux documents

32318

Supelco

Diethylenetriaminepentaacetic acid

for complexometry, ≥99.0%

Synonyme(s) :

(Carboxymethylimino)bis(ethylenenitrilo)tetraacetic acid, N,N-Bis(2-[bis(carboxymethyl)amino]ethyl)glycine, DETAPAC, DTPA, Penta(carboxymethyl)diethylenetriamine, Pentetic acid

Se connecterpour consulter vos tarifs contractuels et ceux de votre entreprise/organisme


About This Item

Formule linéaire :
[(HOOCCH2)2NCH2CH2]2NCH2COOH
Numéro CAS:
Poids moléculaire :
393.35
Beilstein:
1810219
Numéro CE :
Numéro MDL:
Code UNSPSC :
12000000
ID de substance PubChem :

Qualité

for complexometry

Essai

≥99.0% (KT)
≥99.0%

Résidus de calcination

≤0.05% (as SO4)

Perte

≤0.1% loss on drying, 20 °C (HV)

Pf

219-220 °C (lit.)

Solubilité

0.1 M NaOH: 0.1 M at 20 °C, clear, colorless

Chaîne SMILES 

OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CCN(CC(O)=O)CC(O)=O

InChI

1S/C14H23N3O10/c18-10(19)5-15(1-3-16(6-11(20)21)7-12(22)23)2-4-17(8-13(24)25)9-14(26)27/h1-9H2,(H,18,19)(H,20,21)(H,22,23)(H,24,25)(H,26,27)

Clé InChI

QPCDCPDFJACHGM-UHFFFAOYSA-N

Vous recherchez des produits similaires ? Visite Guide de comparaison des produits

Description générale

Visit our Titration Center to learn more.

Autres remarques

Preparation of the complexes: Na2Fe(DTPA)], Na2(Cr(DTPA)) and H2[Gd(DTPA)], which have been found to function extremely well as water soluble paramagnetic relaxation reagents for 13C NMR spectroscopy

Pictogrammes

Health hazardExclamation mark

Mention d'avertissement

Danger

Mentions de danger

Classification des risques

Acute Tox. 4 Inhalation - Eye Irrit. 2 - Repr. 1B - STOT RE 2 Inhalation

Organes cibles

Respiratory Tract

Code de la classe de stockage

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

Classe de danger pour l'eau (WGK)

WGK 2

Point d'éclair (°F)

392.0 °F - closed cup

Point d'éclair (°C)

200 °C - closed cup

Équipement de protection individuelle

dust mask type N95 (US), Eyeshields, Gloves


Faites votre choix parmi les versions les plus récentes :

Certificats d'analyse (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

Si vous avez besoin d'assistance, veuillez contacter Service Clients

Déjà en possession de ce produit ?

Retrouvez la documentation relative aux produits que vous avez récemment achetés dans la Bibliothèque de documents.

Consulter la Bibliothèque de documents

T.J. Wenzel et al.
Analytical Chemistry, 54, 615-615 (1982)
Sébastien Leguay et al.
Inorganic chemistry, 51(23), 12638-12649 (2012-11-17)
Complexation of trivalent actinides with DTPA (diethylenetriamine pentaacetic acid) was studied as a function of pcH and temperature in (Na,H)Cl medium of 0.1 M ionic strength. Formation constants of both complexes AnHDTPA(-) and AnDTPA(2-) (where An stands for Am, Cm
Cerebral perfusion scintigraphy study as confirmation test of brain death in the process of organ donation for transplant.
J Banzo et al.
Revista espanola de medicina nuclear e imagen molecular, 31(5), 278-285 (2012-10-17)
Katsuhiko Sueda et al.
Health physics, 105(2), 208-214 (2013-06-27)
Diethylenetriaminepentaacetic acid (DTPA) is a chelating agent that is used to facilitate the elimination of radionuclides such as americium from contaminated individuals. Its primary site of action is in the blood, where it competes with various biological ligands, including transferrin
Boyd L O'Dell et al.
Biological trace element research, 152(1), 98-104 (2013-01-08)
Zinc deficiency in 3T3 cells induced by the use of diethylenetriaminepentaacetate (DTPA) has been shown to impair calcium entry associated with failure of proliferation when the cells are stimulated with polypeptide growth factors (GF). These functions of zinc have been

Notre équipe de scientifiques dispose d'une expérience dans tous les secteurs de la recherche, notamment en sciences de la vie, science des matériaux, synthèse chimique, chromatographie, analyse et dans de nombreux autres domaines..

Contacter notre Service technique