Principaux documents
Z285447
Sigma-Aldrich
Aldrich® straight vacuum-distilling adapter
joint: ST/NS 24/40
1 EA
90,90 €
90,90 €
Check Cart for Availability
Changer de vue
1 EA
90,90 €
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
90,90 €
Check Cart for Availability
Joint
joint: ST/NS24/40
Catégories apparentées
Description générale
With side hose connection and drip-tip. Improves efficiency of evaporation of solvents under reduced pressure when placed between the condenser of a rotary evaporator and a chilled receiving flask. Plug the condenser vacuum take-off exit opposite the receiving flask to eliminate path competition and direct all distillate to the chilled collection flask. SJ 35/25 adapter fits standard Büchi rotary evaporator glassware.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC