Principaux documents
GX308070516
Supelco
Quartz Torch
with 1.0 mm Injector
About This Item
Code UNSPSC :
41115710
Nomenclature NACRES :
SB.32
Qualité
for Agilent7500
with 1.0 mm Injector
Fabricant/nom de marque
(30-807-0516)
Compatibilité
Agilent/HP 7500
Description générale
Choose GAZ-0604 GazFit Connectors for easy connection of gas tubes to torch side arms