Principaux documents
CFO08NC
Cannon-Fenske Opaque Viscometer for Transparent and Opaque Liquids
Size 350, Nom. Constant 0.5, Range 100 to 500 cSt, (non certified)
About This Item
Code UNSPSC :
41116107
Nomenclature NACRES :
NA.24
Fabricant/nom de marque
Paragon Scientific Ltd
Application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
Description générale
This is the non certified version of this viscometer. If you wish to order a certifed viscometer, please refer to the corresponding product CFO08UKC