Principaux documents
96100023
Millipore
Vantage L Adjuster Cartridge Housing Kit
About This Item
Code UNSPSC :
23151816
eCl@ss :
32011301
Matériaux
glass filled polypropylene surface (exterior)
Fabricant/nom de marque
Vantage
Technique(s)
protein purification: suitable
Diamètre
1.1 cm (0.4 in.)