Key Documents
Z763292
Sigma-Aldrich
Aldrich® Sure/Seal septa, EPDM rubber with PTFE face
pkg of 10 ea
Synonyme(s) :
Sure/seal bottles, Sure/seal septa
About This Item
Code UNSPSC :
41121800
Conditionnement
pkg of 10 ea
Description générale
Dimensions: 26.2 mm diameter x 5 mm thick at center x 1.4 mm thick at rim.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Seal is a trademark of Sigma-Aldrich Co. LLC
Pièce de rechange