Principaux documents
Z748285
Sigma-Aldrich
Load flask for Aldrich® Kugelrohr apparatus
capacity 2000 mL, joint: ST/NS 24/40
Synonyme(s) :
distillation apparatus, distilling apparatus, fractional distillation apparatus, glass distillation apparatus, kugelrohr apparatus, kugelrohr distillation, short path distillation apparatus, vacuum distillation apparatus
1 EA
302,00 €
302,00 €
Check Cart for Availability
Changer de vue
1 EA
302,00 €
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
302,00 €
Check Cart for Availability
Matériaux
glass
single-neck
spherical
Joint
joint: ST/NS24/40
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
case of 1
Fabricant/nom de marque
Sigma-Aldrich
Capacité
2000 mL
Description générale
Heavy wall glass flask with solid support that attaches to Kugelrohr motor drive coupling. Fill load flask one-third full with distillable material.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC