Principaux documents
Z742268
DURAN® TILT GL 56 self-adhesive identification label for screw cap
diam. 40 mm, white polyester, pack of 60 ea (6 labels per sheet)
Synonyme(s) :
294015604
About This Item
Code UNSPSC :
55121611
Nomenclature NACRES :
NB.17
Matériaux
white polyester
Conditionnement
pack of 60 ea (6 labels per sheet)
Fabricant/nom de marque
DURAN® 294015604
Diamètre
40 mm
Description générale
Careful labeling is very important to prevent mix ups and mistakes. The GL 56 self-adhesive cap labels can be used to clearly indicate the separate bottles of media for each cell line, preventing possible cross-contamination.
Caractéristiques et avantages
• Manufactured from polyester• Can be used to clearly indicate the separate bottles of media for each cell line, preventing possible cross-contamination• High temperature performance range makes them ideal for use in fridges, freezers, autoclaves, incubators, and water baths• The labels have a high adhesive strength they are peelable and easy to remove which safes time for cleaning
Informations légales
DURAN is a registered trademark of DWK Life Sciences
À utiliser avec