Principaux documents
Z683205
ALNOCHROMIX safety vent caps
fits 1 gal or 2.5 L glass bottles
3 EA
98,00 €
98,00 €
Check Cart for Availability
Changer de vue
3 EA
98,00 €
About This Item
Code UNSPSC :
41103200
Nomenclature NACRES :
NB.35
98,00 €
Check Cart for Availability
Fabricant/nom de marque
Alconox, Inc. 2533
Description générale
When storing ALNOCHROMIX solution in the Sulfuric Acid glass bottle (1 gallon or 2.5 liter), our Safety Vent Cap will prevent pressure build-up by permitting gases to escape automatically as internal pressure increases.
Informations légales
ALNOCHROMIX is a trademark of Alconox, Inc.