Principaux documents
Z674486
Duran® DG safety cap accessories
venting valve for DG safety caps, PC
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Fabricant/nom de marque
DURAN® 1168430
Description générale
DG safety waste caps are ideal for safely collecting and storing liquid laboratory waste. With a choice of different connections and activated carbon filters, they increase safety for laboratory technicians.
  • Drip-free collection of used or contaminated media
  • Charcoal filters prevent harmful gas emissions
For use with Z674451
Informations légales
DURAN is a registered trademark of DWK Life Sciences