Principaux documents
Z509027
Sigma-Aldrich
AldraSORB water trapping packets
volume 1,000-4,000 mL, large
Synonyme(s) :
Humidity Control Packets
5 EA
122,00 €
25 EA
453,00 €
122,00 €
Check Cart for Availability
Changer de vue
5 EA
122,00 €
25 EA
453,00 €
About This Item
Code UNSPSC :
24141516
Nomenclature NACRES :
NB.43
122,00 €
Check Cart for Availability
Capacité
1000-4000 mL
Volume
1,000-4,000 mL
Poids
20 g
Description générale
The AldraSorb packets are made from an ultra-high molecular weight polyethylene, electronic grade, hydrophobic membrane that contains pharmaceutical quality molecular sieves. The full filtration capability of the molecular particulates is maintained and the surrounding solvent is not compromised. Packets are vacuum sealed.
  • Maintains low levels of water impurities for up to 3 months
  • Ideal for solid-phase and solution-phase peptide synthesis
  • Effectively dries solvents to below 50 ppm
Note: These traps are not recommended for solvents having more than 0.05% or 500 ppm of water. They should not be used in acidic solvents.
Informations légales
AldraSORB is a trademark of Sigma-Aldrich Co. LLC