Principaux documents
Z421561
Sigma-Aldrich
Aldrich® dual-action NMR tube cleaner
joint: ST/NS 24/40, single position
1 EA
653,80 €
653,80 €
Tarif934,00 €Économiser 30 %
Check Cart for Availability
Changer de vue
1 EA
653,80 €
About This Item
Code UNSPSC :
47131800
Nomenclature NACRES :
SB.32
653,80 €
Tarif934,00 €Économiser 30 %
Check Cart for Availability
Joint
joint: ST/NS24/40
Description générale
  • Washes tubes inside and out simultaneously
  • Specially designed to prevent accidental breakage of tubes
  • Optional 1,000 mL reservoir flasks
  • Highly efficient solvent jets improve cleaning and reduce solvent usage
  • For 7 or 8 in. L, 5 mm NMR tubes
Pear-shaped filter flask sold separately.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC