Principaux documents
Z302198
Tyvek® Pro.Tech total body coveralls
size XXL
About This Item
Code UNSPSC :
41100000
Taille
XXL
Fabricant/nom de marque
Tyvek
Disponibilité
not available in USA and Canada
Description générale
With full length zipper, elasticated wrist holes, integral elasticated hood and overshoes for protection in controlled, clean environments. Provides CE type 5 and Type 6 (particle and limited splash) protection.
Informations légales
Tyvek is a registered trademark of E. I. du Pont de Nemours and Company
Remplacé(e)(s) par