Principaux documents
Z286699
Microcells for WILMAD® 5 mm microcell assemblies
capillary, volume 8 μL
Synonyme(s) :
Wilmad nmr tubes, nmr sample tubes, nmr tubes
About This Item
Code UNSPSC :
41121705
Nomenclature NACRES :
SB.32
Conditionnement
pack of 10 ea
Fabricant/nom de marque
Wilmad 529-D
Volume
8 μL
Description générale
Microcells may be used interchangeably in the PTFE holder of the Microcell Assembly and can give the flexibility of taking NMR spectra of small samples.
Informations légales
Wilmad is a registered trademark of SP Industries